tert-Butyl disulfide, 98%
tert-Butyl disulfide, 98%
tert-Butyl disulfide, 98%
Thermo Scientific Chemicals

tert-Butyl disulfide, 98%

CAS: 110-06-5 | C8H18S2 | 178.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 173620025
Price (EUR)
798,00
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
798,00
Each
Chemical Identifiers
CAS110-06-5
IUPAC Name2-(tert-butyldisulfanyl)-2-methylpropane
Molecular FormulaC8H18S2
InChI KeyBKCNDTDWDGQHSD-UHFFFAOYSA-N
SMILESCC(C)(C)SSC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive index1.4890 to 1.4910 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only