tert-Butyl disulfide, 98%
tert-Butyl disulfide, 98%
tert-Butyl disulfide, 98%
Thermo Scientific Chemicals

tert-Butyl disulfide, 98%

CAS: 110-06-5 | C8H18S2 | 178.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 173622500
Price (EUR)
92,65
Online exclusive
103,00
Save 10,35 (10%)
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
92,65
Online exclusive
103,00
Save 10,35 (10%)
Each
Specifications
Assay Percent Range97.5% min. (GC)
Beilstein01,IV,1638
CAS110-06-5
Chemical Name or Materialtert-Butyl disulfide
ColorYellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only