2-Aminobenzimidazole, 99+%
2-Aminobenzimidazole, 99+%
2-Aminobenzimidazole, 99+%
Thermo Scientific Chemicals

2-Aminobenzimidazole, 99+%

CAS: 934-32-7 | C7H7N3 | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 174020250
Price (EUR)
87,30
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
87,30
Each
Chemical Identifiers
CAS934-32-7
IUPAC Name1H-1,3-benzodiazol-2-amine
Molecular FormulaC7H7N3
InChI KeyJWYUFVNJZUSCSM-UHFFFAOYSA-N
SMILESNC1=NC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or flakes
Infrared spectrumConforms
Appearance (Color)Yellow to beige to light brown
AppearanceMay darken on storage
Titration with HClO4>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only