Methyl propiolate, 99%
Methyl propiolate, 99%
Methyl propiolate, 99%
Thermo Scientific Chemicals

Methyl propiolate, 99%

CAS: 922-67-8 | C4H4O2 | 84.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 174040500
Price (EUR)
314,00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
314,00
Each
Chemical Identifiers
CAS922-67-8
IUPAC Namemethyl prop-2-ynoate
Molecular FormulaC4H4O2
InChI KeyIMAKHNTVDGLIRY-UHFFFAOYSA-N
SMILESCOC(=O)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless
Refractive index1.4070 to 1.4090 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only