Nitrosonium tetrafluoroborate, 97%
Nitrosonium tetrafluoroborate, 97%
Nitrosonium tetrafluoroborate, 97%
Thermo Scientific Chemicals

Nitrosonium tetrafluoroborate, 97%

CAS: 14635-75-7 | BF4HNO2 | 133.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number 174391000
Price (EUR)
590,00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
590,00
Each
Chemical Identifiers
CAS14635-75-7
IUPAC Namedioxoamine; tetrafluoroboranuide
Molecular FormulaBF4HNO2
InChI KeyXYRQURYIJKJVSF-UHFFFAOYSA-N
SMILES[O-][NH+]=O.F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay>=96.0 % (ex F)
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only