Chloroform-d, for NMR, 99.6+ atom % D
Chloroform-d, for NMR, 99.6+ atom % D
Chloroform-d, for NMR, 99.6+ atom % D
Thermo Scientific Chemicals

Chloroform-d, for NMR, 99.6+ atom % D

CAS: 865-49-6 | CHCl3 | 120.375 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Catalog number 174881000
Price (EUR)
-
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Beilstein01, IV, 54
Boiling Point60.0°C
CAS865-49-6
ChEBICHEBI:85365
Chemical Name or MaterialChloroform-d
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only