Silver trifluoromethanesulfonate, 99+%
Silver trifluoromethanesulfonate, 99+%
Silver trifluoromethanesulfonate, 99+%
Thermo Scientific Chemicals

Silver trifluoromethanesulfonate, 99+%

CAS: 2923-28-6 | CAgF3O3S | 256.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 175200100
Price (EUR)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2923-28-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Elemental Analysis>=99.0 %
Appearance (Color)White to light beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only