3-Quinolinecarboxaldehyde, 97+%
3-Quinolinecarboxaldehyde, 97+%
3-Quinolinecarboxaldehyde, 97+%
Thermo Scientific Chemicals

3-Quinolinecarboxaldehyde, 97+%

CAS: 13669-42-6 | C10H7NO | 157.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 175690010
Price (EUR)
101,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
101,00
Each
Chemical Identifiers
CAS13669-42-6
IUPAC Namequinoline-3-carbaldehyde
Molecular FormulaC10H7NO
InChI KeyRYGIHSLRMNXWCN-UHFFFAOYSA-N
SMILESO=CC1=CN=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Melting point66°C to 69°C
Infrared spectrumConforms
GC>=97.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only