Pyruvic aldehyde, 30-45 wt% solution in water
Pyruvic aldehyde, 30-45 wt% solution in water
Pyruvic aldehyde, 30-45 wt% solution in water
Thermo Scientific Chemicals

Pyruvic aldehyde, 30-45 wt% solution in water

CAS: 78-98-8 | C3H4O2 | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 175791000
Price (EUR)
65,65
キャンペーン価格
72,80
Save 7,15 (10%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
65,65
キャンペーン価格
72,80
Save 7,15 (10%)
Each
Chemical Identifiers
CAS78-98-8
IUPAC Name2-oxopropanal
Molecular FormulaC3H4O2
InChI KeyAIJULSRZWUXGPQ-UHFFFAOYSA-N
SMILESCC(=O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear yellow to brown
Titration with NaOH30 to 45 %
Refractive index1.3910 to 1.4200 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only