Diphenylcyclopropenone, 98%
Diphenylcyclopropenone, 98%
Diphenylcyclopropenone, 98%
Thermo Scientific Chemicals

Diphenylcyclopropenone, 98%

CAS: 886-38-4 | C15H10O | 206.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 175820010
Price (EUR)
62,50
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
62,50
Each
Chemical Identifiers
CAS886-38-4
IUPAC Namediphenylcycloprop-2-en-1-one
Molecular FormulaC15H10O
InChI KeyHCIBTBXNLVOFER-UHFFFAOYSA-N
SMILESO=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Almost white to light orange-pink to beige
Melting point117°C to 121°C
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only