4-Amino-1-butanol, 98%
4-Amino-1-butanol, 98%
4-Amino-1-butanol, 98%
Thermo Scientific Chemicals

4-Amino-1-butanol, 98%

CAS: 13325-10-5 | C4H11NO | 89.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 176350050
Price (EUR)
82,10
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
82,10
Each
Chemical Identifiers
CAS13325-10-5
IUPAC Name4-hydroxybutan-1-aminium
Molecular FormulaC4H12NO
InChI KeyBLFRQYKZFKYQLO-UHFFFAOYSA-O
SMILES[NH3+]CCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4580 to 1.4630 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only