Dimethyl terephthalate, 99%
Dimethyl terephthalate, 99%
Dimethyl terephthalate, 99%
Thermo Scientific Chemicals

Dimethyl terephthalate, 99%

CAS: 120-61-6 | C10H10O4 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 180580250
Price (EUR)
19,70
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
19,70
Each
Chemical Identifiers
CAS120-61-6
IUPAC Name1,4-dimethyl benzene-1,4-dicarboxylate
Molecular FormulaC10H10O4
InChI KeyWOZVHXUHUFLZGK-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=C1)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point140°C to 143°C
Acid value=<0.03 mg KOH/g
Appearance (Color)White
Appearance (Form)Flakes and/or pellets
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only