Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000
Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000
Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000
Thermo Scientific Chemicals

Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000

Indicator | CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
Catalog number 183380010
Price (EUR)
152,65
Online exclusive
169,00
Save 16,35 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
152,65
Online exclusive
169,00
Save 16,35 (10%)
Each
Chemical Identifiers
CAS9002-89-5
Molecular Formula(C2H4O)n
SMILESOC(-*)C-*
MDL NumberMFCD00081922
SpecificationsSpecification SheetSpecification Sheet
Viscosity9 to 11 cP (4 % in water)
Ash=<1.2 % (as Na2O)
Appearance (Color)White to cream
Appearance (Form)Granular powder
Degree of hydrolysis98.0 to 98.8 mol %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only