4-Phenyl-2-butanol, 97%
4-Phenyl-2-butanol, 97%
4-Phenyl-2-butanol, 97%
Thermo Scientific Chemicals

4-Phenyl-2-butanol, 97%

CAS: 2344-70-9 | C10H14O | 150.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1834000505 gGlass bottle
Catalog number 183400050
Price (EUR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Assay Percent Range96% min. (GC)
Beilstein06,522
CAS2344-70-9
Chemical Name or Material4-Phenyl-2-butanol
Density0.9800g/mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only