Methylmagnesium bromide, 3M solution in diethyl ether, AcroSeal™
Methylmagnesium bromide, 3M solution in diethyl ether, AcroSeal™
Methylmagnesium bromide, 3M solution in diethyl ether, AcroSeal™
Thermo Scientific Chemicals

Methylmagnesium bromide, 3M solution in diethyl ether, AcroSeal™

CAS: 75-16-1 | CH3BrMg | 119.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 183548000
Price (EUR)
226,00
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
226,00
Each
Chemical Identifiers
CAS75-16-1
IUPAC Namebromo(methyl)magnesium
Molecular FormulaCH3BrMg
InChI KeyAVFUHBJCUUTGCD-UHFFFAOYSA-M
SMILESC[Mg]Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to hazy grey to brown to black
Appearance (Form)Solution
AppearanceMay develop some turbidity or precipitate
Titration Grignard2.85 to 3.20 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only