2-Sulfobenzoic acid cyclic anhydride, 95%
2-Sulfobenzoic acid cyclic anhydride, 95%
2-Sulfobenzoic acid cyclic anhydride, 95%
Thermo Scientific Chemicals

2-Sulfobenzoic acid cyclic anhydride, 95%

CAS: 81-08-3 | C7H4O4S | 184.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 184750250
Price (EUR)
67,30
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
67,30
Each
Chemical Identifiers
CAS81-08-3
IUPAC Name3H-2,1λ⁶-benzoxathiole-1,1,3-trione
Molecular FormulaC7H4O4S
InChI KeyNCYNKWQXFADUOZ-UHFFFAOYSA-N
SMILESO=C1OS(=O)(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Titration by coupling>=94 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only