3-Thiophenecarboxaldehyde, 98%
3-Thiophenecarboxaldehyde, 98%
3-Thiophenecarboxaldehyde, 98%
Thermo Scientific Chemicals

3-Thiophenecarboxaldehyde, 98%

CAS: 498-62-4 | C5H4OS | 112.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 187980250
Price (EUR)
148,00
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
148,00
Each
Chemical Identifiers
CAS498-62-4
IUPAC Namethiophene-3-carbaldehyde
Molecular FormulaC5H4OS
InChI KeyRBIGKSZIQCTIJF-UHFFFAOYSA-N
SMILESO=CC1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5820 to 1.5860 (20°C, 589 nm)
Appearance (Color)Clear yellow to light brown
AppearanceMay darken on storage
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only