Di-tert-butyl dicarbonate, 99%
Di-tert-butyl dicarbonate, 99%
Di-tert-butyl dicarbonate, 99%
Thermo Scientific Chemicals

Di-tert-butyl dicarbonate, 99%

CAS: 24424-99-5 | C10H18O5 | 218.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
50 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 194671000
Price (EUR)
199,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
199,00
Each
Chemical Identifiers
CAS24424-99-5
IUPAC Namedi-tert-butyl dicarbonate
Molecular FormulaC10H18O5
InChI KeyDYHSDKLCOJIUFX-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)OC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Low melting solid
Infrared spectrumConforms
Titration by coupling98.5 to 101.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only