Platinum(II) chloride, 73% Pt
Platinum(II) chloride, 73% Pt
Platinum(II) chloride, 73% Pt
Thermo Scientific Chemicals

Platinum(II) chloride, 73% Pt

CAS: 10025-65-7 | Cl2Pt | 265.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number 195380010
Price (EUR)
177,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
177,00
Each
Chemical Identifiers
CAS10025-65-7
IUPAC Nameplatinum(2+) dichloride
Molecular FormulaCl2Pt
InChI KeyCLSUSRZJUQMOHH-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pt++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Grey-green to brown
Assay>=71.8 % (Pt)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only