Iodine monochloride, 98%
Iodine monochloride, 98%
Iodine monochloride, 98%
Thermo Scientific Chemicals

Iodine monochloride, 98%

CAS: 7790-99-0 | ClI | 162.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 196965000
Price (EUR)
239,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
239,00
Each
Specifications
Assay Percent Range97.5% min. (Iodometry)
Boiling Point97.0°C
CAS7790-99-0
Chemical Name or MaterialIodine monochloride
ColorRed to Brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only