Titanium(IV) chloride, 99.9%
Titanium(IV) chloride, 99.9%
Titanium(IV) chloride, 99.9%
Thermo Scientific Chemicals

Titanium(IV) chloride, 99.9%

CAS: 7550-45-0 | Cl4Ti | 189.667 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 197230010
Price (EUR)
353,65
온라인 행사
393,00
Save 39,35 (10%)
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
353,65
온라인 행사
393,00
Save 39,35 (10%)
Each
Chemical Identifiers
CAS7550-45-0
IUPAC Nametitanium(4+) tetrachloride
Molecular FormulaCl4Ti
InChI KeyXJDNKRIXUMDJCW-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow
Aluminium (Al)=<2 ppm
Iron (Fe)=<2 ppm
Appearance (Form)Liquid
Assay>=99.85 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only