Copper(II) sulfate pentahydrate, 99+%, for analysis
Copper(II) sulfate pentahydrate, 99+%, for analysis
Copper(II) sulfate pentahydrate, 99+%, for analysis
Copper(II) sulfate pentahydrate, 99+%, for analysis
Thermo Scientific Chemicals

Copper(II) sulfate pentahydrate, 99+%, for analysis

CAS: 7758-99-8 | CuH10O9S | 249.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 197720010
Price (EUR)
54,65
Online Exclusive
60,30
Save 5,65 (9%)
Each
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
54,65
Online Exclusive
60,30
Save 5,65 (9%)
Each
Add to cart
Chemical Identifiers
CAS7758-99-8
IUPAC Namecopper(2+) pentahydrate sulfate
Molecular FormulaCuH10O9S
InChI KeyJZCCFEFSEZPSOG-UHFFFAOYSA-L
SMILESO.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Chloride (Cl)=<10 ppm
Nickel (Ni)=<20 ppm
Iron (Fe)=<50 ppm
Total nitrogen (as N)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only