Potassium periodate, 99.5%, extra pure
Potassium periodate, 99.5%, extra pure
Potassium periodate, 99.5%, extra pure
Potassium periodate, 99.5%, extra pure
Thermo Scientific Chemicals

Potassium periodate, 99.5%, extra pure

CAS: 7790-21-8 | IKO4 | 230 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 198391000
Price (EUR)
92,40
Each
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
92,40
Each
Add to cart
Chemical Identifiers
CAS7790-21-8
IUPAC Namepotassium periodate
Molecular FormulaIKO4
InChI KeyFJVZDOGVDJCCCR-UHFFFAOYSA-M
SMILES[K+].[O-][I](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Manganese (Mn)=<1 ppm
Titration Iodometric>=99.4 %
Other halogens (as Cl)=<0.01 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only