Trimethylsilyl cyanide, 98%
Trimethylsilyl cyanide, 98%
Trimethylsilyl cyanide, 98%
Thermo Scientific Chemicals

Trimethylsilyl cyanide, 98%

CAS: 7677-24-9 | C4H9NSi | 99.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 199560250
Price (EUR)
163,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
163,00
Each
Chemical Identifiers
CAS7677-24-9
IUPAC Nametrimethylsilanecarbonitrile
Molecular FormulaC4H9NSi
InChI KeyLEIMLDGFXIOXMT-UHFFFAOYSA-N
SMILESC[Si](C)(C)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow to yellow-brown
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.3900 to 1.3930 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only