Selenium(IV) oxide, 99.8%, (trace metal basis)
Selenium(IV) oxide, 99.8%, (trace metal basis)
Selenium(IV) oxide, 99.8%, (trace metal basis)
Thermo Scientific Chemicals

Selenium(IV) oxide, 99.8%, (trace metal basis)

CAS: 7446-08-4 | O2Se | 110.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 200000025
Price (EUR)
1.443,00
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
1.443,00
Each
Chemical Identifiers
CAS7446-08-4
IUPAC Name(oxo-λ⁴-selanylidene)oxidane
Molecular FormulaO2Se
InChI KeyJPJALAQPGMAKDF-UHFFFAOYSA-N
SMILESO=[Se]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Copper (Cu)=<10 ppm
Cadmium (Cd)=<5 ppm
Nickel (Ni)=<10 ppm
Appearance (Color)White to light beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only