Nalgene™ Rectangular Amber HDPE Bottles
Nalgene™ Rectangular Amber HDPE Bottles
Nalgene™ Rectangular Amber HDPE Bottles
Nalgene™ Rectangular Amber HDPE Bottles
Thermo Scientific™

Nalgene™ Rectangular Amber HDPE Bottles

Protect light-sensitive liquids or dry materials from exposure with these space-efficient, amber HDPE bottles, featuring sturdy walls and a wide mouth for easy filling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
2009-0064PK
also known as 2009-0064
2 L63 mmPack of 4
2009-0004125 mL28 mmCase of 72
2009-0004PK
also known as 2009-0004
125 mL28 mmPack of 12
2009-0008250 mL38 mmCase of 72
2009-0008PK
also known as 2009-0008
250 mL38 mmPack of 12
2009-0016500 mL48 mmCase of 48
2009-0016PK
also known as 2009-0016
500 mL48 mmPack of 12
2009-00321 L53 mmCase of 24
2009-0032PK
also known as 2009-0032
1 L53 mmPack of 6
2009-00642 L63 mmCase of 12
Catalog number 2009-0064PK
also known as 2009-0064
Price (EUR)
85,80
Pack of 4
Capacity (Metric):
2 L
Closure Size:
63 mm
Unit Size:
Pack of 4
Price (EUR)
85,80
Pack of 4

Protect light-sensitive liquids or dry materials from exposure. Thermo Scientific™ Nalgene™ Rectangular Amber HDPE Bottles take up less shelf space than traditional round bottles. These high-density, polyethylene bottles have a sturdy wall construction with straight sides and a wide mouth for easy filling. Excellent chemical resistance to most acids, bases and alcohols makes this container great for sampling, shipping or storing liquids or dry materials that are sensitive to UV light.

  • Amber polypropylene screw closure
  • Amber HDPE reduces UV light transmissions to protect light-sensitive liquids
  • Meets U.S. Pharmacopoeia 23 light transmission requirements
  • Rectangular shape is great for space-saving use in the freezer to -100°C (-148°F)
  • Excellent chemical resistance to most acids, bases and alcohols make these excellent containers to protect your valuable samples
  • Linerless caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
.

Recycling Information
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity64 oz., 2000 mL
Closure ColorOpaque Amber
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size63 mm
Closure TypeScrew Cap
DescriptionAmber Wide-Mouth Rectangular Bottle
Diameter (English) Inner Neck2 in.
Diameter (Metric)152.4 mm
Diameter (Metric) Inner Neck51 mm
Height (English)9.5 in.
Height (Metric)242 mm
Length (Metric)152 mm
No. per Case12
No. per Pack4
Through the Closure DispensingNo
Width (Metric)84 mm
Capacity (English)64 oz.
Capacity (Metric)2 L
Certifications/ComplianceMeet U.S. Pharmacopeia XXIII requirements for maximum light transmission.
Closure IncludedYes
ColorAmber
GraduatedNo
MaterialHDPE
MouthWide
Product TypeWide Mouth Bottle
ShapeRectangular
SterilityNon-sterile
Unit SizePack of 4

Frequently asked questions (FAQs)

Why are your Nalgene containers and caps always different materials?

In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.