Pararosaniline base
Pararosaniline base
Pararosaniline base
Thermo Scientific Chemicals

Pararosaniline base

CAS: 467-62-9 | C19H19N3O | 305.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 201550500
Price (EUR)
162,65
Online Exclusive
180,00
Save 17,35 (10%)
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
162,65
Online Exclusive
180,00
Save 17,35 (10%)
Each
Chemical Identifiers
CAS467-62-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Purple
Appearance (Form)Crystalline powder
Infrared spectrumConforms
UV spectrumComplies
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only