Amberlite™ XAD-4
Amberlite™ XAD-4
Amberlite™ XAD-4
Thermo Scientific Chemicals

Amberlite™ XAD-4

CAS: 9003-69-4 | C10H10
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 202231000
Price (EUR)
52,20
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
52,20
Each
Chemical Identifiers
CAS9003-69-4
MDL NumberMFCD00132704
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Moisture holding capacity54.0 to 60.0 %
Uniformity coefficient=<2.0
Particle size=<5.0 % (< 0.300 mm)
Harmonic mean size0.490 to 0.690 mm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only