Amberlite™ IRA-900, Cl-form, ion-exchange resin
Amberlite™ IRA-900, Cl-form, ion-exchange resin
Amberlite™ IRA-900, Cl-form, ion-exchange resin
Thermo Scientific Chemicals

Amberlite™ IRA-900, Cl-form, ion-exchange resin

CAS: 9050-97-9 | C4H12ClN | 109.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 202310025
Price (EUR)
203,65
Online exclusive
226,00
Save 22,35 (10%)
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
203,65
Online exclusive
226,00
Save 22,35 (10%)
Each
Chemical Identifiers
CAS9050-97-9
MDL NumberMFCD00132712
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Beads
Total volume capacity>=1.0 eq/l (Cl-)
Particle size=<3.0 % (> 1.180 mm)
Appearance (Color)Cream to beige to light brown
Uniformity coefficient=<1.8
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only