Potassium chromate, 99.5%, for analysis
Potassium chromate, 99.5%, for analysis
Potassium chromate, 99.5%, for analysis
Thermo Scientific Chemicals

Potassium chromate, 99.5%, for analysis

CAS: 7789-00-6 | CrK2O4 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 202345000
Price (EUR)
108,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
108,00
Each
Chemical Identifiers
CAS7789-00-6
IUPAC Namedipotassium dioxochromiumbis(olate)
Molecular FormulaCrK2O4
InChI KeyXMXNVYPJWBTAHN-UHFFFAOYSA-N
SMILES[K+].[K+].[O-][Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Powder or crystalline powder or crystals
Titration Iodometric>=99.5 %
Chloride (Cl)=<10 ppm
Sulfate (SO4)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysis
RUO – Research Use Only