Hydrogen peroxide, for analysis, 35 wt.% solution in water, stabilized
Hydrogen peroxide, for analysis, 35 wt.% solution in water, stabilized
Hydrogen peroxide, for analysis, 35 wt.% solution in water, stabilized
Thermo Scientific Chemicals

Hydrogen peroxide, for analysis, 35 wt.% solution in water, stabilized

CAS: 7722-84-1 | H2O2 | 34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
1 L
2.5 L
10 L
25 L
Packaging:
View all
Plastic drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 202460100
Price (EUR)
288,00
Each
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
288,00
Each
Chemical Identifiers
CAS7722-84-1
IUPAC Nameperoxol
Molecular FormulaH2O2
InChI KeyMHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILESOO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Heavy metals=<10 ppm
Chloride (Cl)=<2 ppm
Stabilizerabout 0.03% stabilizers
Arsenic (As)=<3 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only