Trichloroacetyl isocyanate, NMR grade
Trichloroacetyl isocyanate, NMR grade
Trichloroacetyl isocyanate, NMR grade
Thermo Scientific Chemicals

Trichloroacetyl isocyanate, NMR grade

CAS: 3019-71-4 | C3Cl3NO2 | 188.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 202900100
Price (EUR)
87,60
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
87,60
Each
Chemical Identifiers
CAS3019-71-4
IUPAC Nametrichloroethanecarbonyl isocyanate
Molecular FormulaC3Cl3NO2
InChI KeyGRNOZCCBOFGDCL-UHFFFAOYSA-N
SMILESClC(Cl)(Cl)C(=O)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4799 to 1.4819 (20°C, 589 nm)
GC>=97 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only