Dichloroisocyanuric acid sodium salt, 98%
Dichloroisocyanuric acid sodium salt, 98%
Dichloroisocyanuric acid sodium salt, 98%
Thermo Scientific Chemicals

Dichloroisocyanuric acid sodium salt, 98%

Dichloroisocyanuric acid sodium salt, >-97.5% (on dry substance), C3HCl2N3NaO3, CAS Number-2893-78-9, sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt, 500g, 98%, White, 220-767-7, 219.95, Danger | CAS: 2893-78-9 | C3HCl2N3NaO3 | 220.949 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Catalog number 204110250
Price (EUR)
39,65
Online Exclusive
44,40
Save 4,75 (11%)
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
39,65
Online Exclusive
44,40
Save 4,75 (11%)
Each
Chemical Identifiers
CAS2893-78-9
IUPAC Namesodium 3,5-dichloro-2,4,6-trioxo-1,3,5-triazinan-1-ide
Molecular FormulaC3Cl2N3NaO3
InChI KeyMSFGZHUJTJBYFA-UHFFFAOYSA-M
SMILES[Na+].ClN1C(=O)[N-]C(=O)N(Cl)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals or granules
Infrared spectrumConforms
Titration with HClO4>=97.5 % (On dry substance)
Loss on drying=<5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagent
RUO – Research Use Only