D-Asparagine monohydrate
Thermo Scientific Chemicals

D-Asparagine monohydrate

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 204270050
Price (EUR)
64,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
64,00
Each
Chemical Identifiers
CAS5794-24-1
IUPAC Name2-amino-3-carbamoylpropanoic acid hydrate
Molecular FormulaC4H10N2O4
InChI KeyRBMGJIZCEWRQES-UHFFFAOYNA-N
SMILESO.NC(CC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Titration Total base98.5 to 101.5 % (on dry substance)
Heavy metals (as Pb)=<20 ppm
Specific optical rotationon dry substance
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only