Isethionic acid, sodium salt, 98%
Isethionic acid, sodium salt, 98%
Isethionic acid, sodium salt, 98%
Thermo Scientific Chemicals

Isethionic acid, sodium salt, 98%

CAS: 1562-00-1 | C2H5NaO4S | 148.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number 205000010
Price (EUR)
66,10
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
66,10
Each
Chemical Identifiers
CAS1562-00-1
IUPAC Namesodium 2-hydroxyethane-1-sulfonate
Molecular FormulaC2H5NaO4S
InChI KeyLADXKQRVAFSPTR-UHFFFAOYSA-M
SMILES[Na+].OCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point190°C to 198°C
Titration after Ion exchange97.5 to 102.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder or powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only