Ethyl methanesulfonate, 99%
Ethyl methanesulfonate, 99%
Ethyl methanesulfonate, 99%
Thermo Scientific Chemicals

Ethyl methanesulfonate, 99%

CAS: 62-50-0 | C3H8O3S | 124.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 205265000
Price (EUR)
2.324,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
2.324,00
Each
Chemical Identifiers
CAS62-50-0
IUPAC Nameethyl methanesulfonate
Molecular FormulaC3H8O3S
InChI KeyPLUBXMRUUVWRLT-UHFFFAOYSA-N
SMILESCCOS(C)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4170 to 1.4190 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only