1-Pentanesulfonic acid, sodium salt, 98+%
1-Pentanesulfonic acid, sodium salt, 98+%
1-Pentanesulfonic acid, sodium salt, 98+%
Thermo Scientific Chemicals

1-Pentanesulfonic acid, sodium salt, 98+%

CAS: 22767-49-3 | C5H11NaO3S | 174.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 206110250
Price (EUR)
131,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
131,00
Each
Chemical Identifiers
CAS22767-49-3
IUPAC Namesodium pentane-1-sulfonate
Molecular FormulaC5H11NaO3S
InChI KeyROBLTDOHDSGGDT-UHFFFAOYSA-M
SMILES[Na+].CCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Loss on drying=<2 % (1 g, 120°C)
Infrared spectrumConforms
Appearance (Form)Powder
Titration after Ion exchange>=98.0 % (on dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only