Sodium molybdate(VI) dihydrate, 99+%
Sodium molybdate(VI) dihydrate, 99+%
Sodium molybdate(VI) dihydrate, 99+%
Thermo Scientific Chemicals

Sodium molybdate(VI) dihydrate, 99+%

CAS: 10102-40-6 | MoNa2O4 | 205.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 206370025
Price (EUR)
755,00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
755,00
Each
Chemical Identifiers
CAS10102-40-6
IUPAC Namedisodium dioxomolybdenumbis(olate) dihydrate
Molecular FormulaH4MoNa2O6
InChI KeyFDEIWTXVNPKYDL-UHFFFAOYSA-N
SMILESO.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=99.0 %
Appearance (Form)Crystalline powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only