Zirconium(IV) chloride, 98%, anhydrous
Zirconium(IV) chloride, 98%, anhydrous
Zirconium(IV) chloride, 98%, anhydrous
Thermo Scientific Chemicals

Zirconium(IV) chloride, 98%, anhydrous

CAS: 10026-11-6 | Cl4Zr | 233.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 206410025
Price (EUR)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10026-11-6
IUPAC Namezirconium(4+) tetrachloride
Molecular FormulaCl4Zr
InChI KeyDUNKXUFBGCUVQW-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Zr+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Chromium (Cr)=<100 ppm
Assay>=97.5 % (ZrCl4)
Aluminium (Al)=<100 ppm
Iron (Fe)=<1000 ppm
Hafnium (Hf)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only