Vanadium(V) oxide, 99.6+%
Vanadium(V) oxide, 99.6+%
Vanadium(V) oxide, 99.6+%
Thermo Scientific Chemicals

Vanadium(V) oxide, 99.6+%

CAS: 1314-62-1 | O5V2 | 181.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 206420010
Price (EUR)
259,00
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
259,00
Each
Chemical Identifiers
CAS1314-62-1
IUPAC Name[(dioxovanadio)oxy]dioxovanadium
Molecular FormulaO5V2
InChI KeyGNTDGMZSJNCJKK-UHFFFAOYSA-N
SMILESO=[V](=O)O[V](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange-brown
Potassium (K)=<200 ppm
Iron (Fe)=<300 ppm
Silicon (Si)=<200 ppm
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only