Tungstic acid, 99+%
Tungstic acid, 99+%
Tungstic acid, 99+%
Tungstic acid, 99+%
Thermo Scientific Chemicals

Tungstic acid, 99+%

CAS: 7783-03-1 | H2O4W | 249.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 207615000
Price (EUR)
225,65
Online Exclusive
250,00
Save 24,35 (10%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
225,65
Online Exclusive
250,00
Save 24,35 (10%)
Each
Chemical Identifiers
CAS7783-03-1
IUPAC Namedioxotungstendiol
Molecular FormulaH2O4W
InChI KeyCMPGARWFYBADJI-UHFFFAOYSA-L
SMILESO[W](O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Assay>=99.0 %
Appearance (Color)Yellow to yellow-green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only