Tolylene 2,6-diisocyanate, 97%
Tolylene 2,6-diisocyanate, 97%
Tolylene 2,6-diisocyanate, 97%
Thermo Scientific Chemicals

Tolylene 2,6-diisocyanate, 97%

CAS: 91-08-7 | C9H6N2O2 | 174.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 208040050
Price (EUR)
195,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
195,00
Each
Chemical Identifiers
CAS91-08-7
IUPAC Name1,3-diisocyanato-2-methylbenzene
Molecular FormulaC9H6N2O2
InChI KeyRUELTTOHQODFPA-UHFFFAOYSA-N
SMILESCC1=C(C=CC=C1N=C=O)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5700 to 1.5720 (20°C, 589 nm)
Appearance (Form)Clear to light hazy liquid or low melting mass
GC>=96.0 %
Appearance (Color)Colorless to white to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only