CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
Thermo Scientific Chemicals

CHES, 98+%, for biochemistry

CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 208180250
Price (EUR)
37,65
Online exclusive
41,40
Save 3,75 (9%)
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
37,65
Online exclusive
41,40
Save 3,75 (9%)
Each
Chemical Identifiers
CAS103-47-9
IUPAC Namesodium 2-(cyclohexylamino)ethane-1-sulfonate
Molecular FormulaC8H16NNaO3S
InChI KeyTXWKJOVDGFRVOG-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)CCNC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 280 nm A: =<0.04
Water=<1 % (K.F.)
Infrared spectrumConforms
UV(0.1 M in water)
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only