Benzyltrimethylammonium chloride, 50 wt% solution in water
Benzyltrimethylammonium chloride, 50 wt% solution in water
Benzyltrimethylammonium chloride, 50 wt% solution in water
Benzyltrimethylammonium chloride, 50 wt% solution in water
Thermo Scientific Chemicals

Benzyltrimethylammonium chloride, 50 wt% solution in water

CAS: 56-93-9 | C10H16ClN | 185.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Catalog number 209381000
Price (EUR)
15,00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
15,00
Each
Add to cart
Chemical Identifiers
CAS56-93-9
IUPAC Namebenzyltrimethylazanium chloride
Molecular FormulaC10H16ClN
InChI KeyKXHPPCXNWTUNSB-UHFFFAOYSA-M
SMILES[Cl-].C[N+](C)(C)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless to light yellow
Titration Argentometric>=49 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only