Silver benzoate, 99%
Silver benzoate, 99%
Silver benzoate, 99%
Thermo Scientific Chemicals

Silver benzoate, 99%

CAS: 532-31-0 | C7H5AgO2 | 228.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 210770100
Price (EUR)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS532-31-0
IUPAC Namesilver(1+) benzoate
Molecular FormulaC7H5AgO2
InChI KeyCLDWGXZGFUNWKB-UHFFFAOYSA-M
SMILES[Ag+].[O-]C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to light grey
Infrared spectrumConforms
Silver (Ag)46.4 to 47.8 %
Appearance (Form)Powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only