Aluminum nitrate nonahydrate, 99.999%, (trace metal basis)
Aluminum nitrate nonahydrate, 99.999%, (trace metal basis)
Aluminum nitrate nonahydrate, 99.999%, (trace metal basis)
Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, 99.999%, (trace metal basis)

CAS: 7784-27-2 | AlN3O9·9H2O | 375.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 212240100
Price (EUR)
70,80
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
70,80
Each
Chemical Identifiers
CAS7784-27-2
IUPAC Namealuminium(3+) nonahydrate trinitrate
Molecular FormulaAlH18N3O18
InChI KeySWCIQHXIXUMHKA-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<10 ppm
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Complexometric6.5 to 7.9 % (Al)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only