Cadmium Carbonate, Pure
Cadmium Carbonate, Pure
Cadmium Carbonate, Pure
Thermo Scientific Chemicals

Cadmium Carbonate, Pure

CAS: 513-78-0 | CCdO3 | 172.42 g/mol
Have Questions?
Catalog NumberQuantity
212322500250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 212322500
Price (EUR)
79,90
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
79,90
Each
Chemical Identifiers
CAS513-78-0
IUPAC Namecadmium(2+) carbonate
Molecular FormulaCCdO3
InChI KeyGKDXQAKPHKQZSC-UHFFFAOYSA-L
SMILES[Cd++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Assay60.0 to 63.0 % (Cd)
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only