Ammonium cerium(IV) nitrate, pure
Ammonium cerium(IV) nitrate, pure
Ammonium cerium(IV) nitrate, pure
Thermo Scientific Chemicals

Ammonium cerium(IV) nitrate, pure

Oxidation reagent | CAS: 16774-21-3 | H8CeN8O18 | 548.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 212360050
Price (EUR)
706,65
Online exclusive
785,00
Save 78,35 (10%)
Each
Quantity:
5 kg
Request bulk or custom format
Price (EUR)
706,65
Online exclusive
785,00
Save 78,35 (10%)
Each
Chemical Identifiers
CAS16774-21-3
IUPAC Nameλ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
Molecular FormulaCeH8N8O18
InChI KeyWIBGOERAEYJBOT-UHFFFAOYSA-N
SMILESN.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Loss on drying=<2 % (110°C)
Appearance (Color)Orange
Titration>=98.5 % (On dry substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only