Potassium fluoride, 99+%, for analysis, Thermo Scientific Chemicals
Potassium fluoride, 99+%, for analysis, Thermo Scientific Chemicals
Potassium fluoride, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium fluoride, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 212600010
Price (EUR)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7789-23-3
IUPAC Namepotassium fluoride
Molecular FormulaFK
InChI KeyNROKBHXJSPEDAR-UHFFFAOYSA-M
SMILES[F-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration99.0 to 101.0 %
Free acid (HF)=<0.1 %
Potassium fluosilicate=<0.1 %
Sulfate (SO4)=<200 ppm
Cadmium (Cd)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only