Methyl (S)-(-)-lactate, 97%
Methyl (S)-(-)-lactate, 97%
Methyl (S)-(-)-lactate, 97%
Thermo Scientific Chemicals

Methyl (S)-(-)-lactate, 97%

CAS: 27871-49-4 | C4H8O3 | 104.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 213060025
Price (EUR)
465,00
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
465,00
Each
Chemical Identifiers
CAS27871-49-4
IUPAC Namemethyl (2S)-2-hydroxypropanoate
Molecular FormulaC4H8O3
InChI KeyLPEKGGXMPWTOCB-VKHMYHEASA-N
SMILESCOC(=O)[C@H](C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Specific optical rotation-8° to -9° (20°C, 589 nm) (neat)
Appearance (Form)Liquid
Free acid=<1.0 % (lactic acid)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only